Assay Details
Gene Symbol: MIR9028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128042.1 | 1 | 93 | 58 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transient receptor potential cation channel subfamily M member 2 | TRPM2 | -3.5kb | Anti-sense | Chr. 26 - 40865559 - 40924941 on NCBI Build EquCab3.0 | |
Downstream | leucine rich repeat containing 3 | LRRC3 | 3.9kb | Anti-sense | Chr. 26 - 40932445 - 40935678 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
