Assay Details
Gene Symbol: MIR9044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128061.1 | 1 | 88 | 90 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chondroitin sulfate N-acetylgalactosaminyltransferase 2 | CSGALNACT2 | -1.0kb | Anti-sense | Chr. 1 - 71543973 - 71585337 on NCBI Build EquCab3.0 | |
Downstream | ret proto-oncogene | RET | 10.7kb | Anti-sense | Chr. 1 - 71597235 - 71651734 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
