Assay Details
Gene Symbol: MIR3548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128081.1 | 1 | 80 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-429 | MIR429 | eca-mir-429 | -1.3kb | Anti-sense | Chr. 2 - 48712423 - 48712489 on NCBI Build EquCab3.0 |
Downstream | microRNA mir-200b | MIR200B | eca-mir-200b | 555.0 | Anti-sense | Chr. 2 - 48714468 - 48714536 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
