Assay Details
Gene Symbol: MIR9070 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128093.1 | 1 | 77 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transmembrane protein 175 | TMEM175 | -26.4kb | Sense | Chr. 3 - 120979355 - 121006653 on NCBI Build EquCab3.0 | |
Downstream | complexin 1 | CPLX1 | 46.6kb | Anti-sense | Chr. 3 - 121079869 - 121116163 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
