Assay Details
Gene Symbol: MIR9074 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128097.1 | 1 | 98 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ankyrin repeat domain 11 | ANKRD11 | -2.4kb | Anti-sense | Chr. 3 - 36424320 - 36637853 on NCBI Build EquCab3.0 | |
Downstream |
SPG7 matrix AAA peptidase subunit, paraplegin |
SPG7 | 12.3kb | Sense | Chr. 3 - 36652624 - 36685180 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
