Assay Details
Gene Symbol: MIR9077 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128100.1 | 1 | 93 | 90 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transmembrane protein 128 | TMEM128 | -32.5kb | Sense | Chr. 3 - 114353639 - 114363550 on NCBI Build EquCab3.0 | |
Downstream | zinc finger and BTB domain containing 49 | ZBTB49 | 1.0kb | Anti-sense | Chr. 3 - 114397234 - 114423321 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
