Assay Details
Gene Symbol: MIR9098 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128125.1 | 1 | 85 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-9107 | MIR9107 | eca-mir-9107 | -26.9kb | Sense | Chr. 11 - 954964 - 955070 on NCBI Build EquCab3.0 |
Downstream | solute carrier family 16 member 3 | SLC16A3 | MCT4 | 4.5kb | Anti-sense | Chr. 11 - 986568 - 998917 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
