Assay Details
Gene Symbol: MIR7035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128134.1 | 1 | 92 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Aly/REF export factor | ALYREF | -644.0 | Sense | Chr. 11 - 1349265 - 1353818 on NCBI Build EquCab3.0 | |
Downstream | Rho GDP dissociation inhibitor alpha | ARHGDIA | 16.2kb | Sense | Chr. 11 - 1370763 - 1375063 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
