Assay Details
Gene Symbol: MIR483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128151.1 | 1 | 74 | 110 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
H19, imprinted maternally expressed transcript (non-protein coding) |
H19 | -103.8kb | Sense | Chr. 12 - 34334581 - 34336980 on NCBI Build EquCab3.0 | |
Downstream | insulin | INS | 25.4kb | Sense | Chr. 12 - 34466340 - 34467800 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
