Assay Details
Gene Symbol: MIR1379 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128155.1 | 1 | 53 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | TATA box-binding protein-associated factor RNA polymerase I subunit A | LOC102147783 | -18.2kb | Anti-sense | Chr. 30 - 12921115 - 12960489 on NCBI Build EquCab3.0 | |
Downstream | solute carrier family 30 member 10 | SLC30A10 | 89.4kb | Sense | Chr. 30 - 13068219 - 13081952 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
