Assay Details
Gene Symbol: MIR9124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128156.1 | 1 | 63 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger protein 281 | ZNF281 | -85.4kb | Sense | Chr. 30 - 28930874 - 28951383 on NCBI Build EquCab3.0 | |
Downstream | kinesin family member 14 | KIF14 | 16.6kb | Sense | Chr. 30 - 29053449 - 29106180 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
