Assay Details
Gene Symbol: MIR9126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128158.1 | 1 | 89 | 108 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tumor protein p53 binding protein 2 | TP53BP2 | Sense | Chr. 30 - 9556530 - 9614337 on NCBI Build EquCab3.0 | ||
Downstream | calpain 2 | CAPN2 | 19.2kb | Anti-sense | Chr. 30 - 9618890 - 9677885 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
