Assay Details
Gene Symbol: MIR7177B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128173.1 | 1 | 53 | 112 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cytosolic arginine sensor for mTORC1 subunit 2 | CASTOR2 | GATSL2 | -2.8mb | Anti-sense | Chr. 13 - 13599726 - 13661530 on NCBI Build EquCab3.0 |
Downstream | polypeptide N-acetylgalactosaminyltransferase 17 | GALNT17 | WBSCR17 | 449.7kb | Sense | Chr. 13 - 16895260 - 17349926 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
