Assay Details
Gene Symbol: MIR507B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128181.1 | 1 | 77 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-508 | MIR508 | eca-mir-508 | -6.1kb | Sense | Chr. X - 120863509 - 120863625 on NCBI Build EquCab3.0 |
Downstream | microRNA mir-509b | MIR509B | eca-mir-509b | 11.1kb | Sense | Chr. X - 120880951 - 120881083 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
