Assay Details
Gene Symbol: MIR2483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128183.1 | 1 | 83 | 104 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SH2 domain containing 1A | SH2D1A | -733.2kb | Anti-sense | Chr. X - 102826898 - 102850830 on NCBI Build EquCab3.0 | |
Downstream | TEX13 family member C | TEX13C | 98.3kb | Anti-sense | Chr. X - 103682565 - 103685993 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
