Assay Details
Gene Symbol: MIR676 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128192.1 | 1 | 76 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | family with sequence similarity 155 member B | FAM155B | -404.3kb | Sense | Chr. X - 54591263 - 54631494 on NCBI Build EquCab3.0 | |
Downstream | acyl-CoA wax alcohol acyltransferase 2 | AWAT2 | 18.5kb | Anti-sense | Chr. X - 55054400 - 55062338 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
