Assay Details
Gene Symbol: MIR509B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128197.1 | 1 | 57 | 105 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-507b | MIR507B | eca-mir-507b | -11.1kb | Sense | Chr. X - 120869739 - 120869875 on NCBI Build EquCab3.0 |
Downstream | microRNA mir-509a | MIR509A |
MIR509, eca-mir-509, eca-mir-509a |
5.5kb | Sense | Chr. X - 120886540 - 120886637 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
