Assay Details
Gene Symbol: MIR2114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128204.1 | 1 | 76 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
heat shock transcription factor, X-linked member 3 |
LOC100069804 | -287.2kb | Sense | Chr. X - 122980903 - 122983151 on NCBI Build EquCab3.0 | |
Downstream | mastermind like domain containing 1 | MAMLD1 | f18 | 126.4kb | Sense | Chr. X - 123396878 - 123521456 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
