Assay Details
Gene Symbol: MIR9144 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128216.1 | 1 | 76 | 94 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cyclin and CBS domain divalent metal cation transport mediator 3 | CNNM3 | -5.6kb | Sense | Chr. 15 - 12043295 - 12059823 on NCBI Build EquCab3.0 | |
Downstream |
lectin, mannose binding 2 like |
LMAN2L | 49.8kb | Anti-sense | Chr. 15 - 12115385 - 12140193 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
