Assay Details
Gene Symbol: MIR9151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128224.1 | 1 | 69 | 94 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | TBC1 domain family member 8 | TBC1D8 | Sense | Chr. 15 - 8554501 - 8667589 on NCBI Build EquCab3.0 | ||
Downstream | ribosomal protein L31 | RPL31 | 73.7kb | Anti-sense | Chr. 15 - 8668000 - 8674208 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
