Assay Details
Gene Symbol: MIR1543 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128230.1 | 1 | 83 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | small nucleolar RNA host gene 4 (non-protein coding) | SNHG4 | Sense | Chr. 14 - 36671555 - 36675518 on NCBI Build EquCab3.0 | ||
Downstream | SIL1 nucleotide exchange factor | SIL1 | 3.6kb | Anti-sense | Chr. 14 - 36675646 - 36929681 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
