Assay Details
| Gene Symbol: C11H17orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362501.4 | XP_003362549.2 | 2216 | 103 | ||
| XM_023652998.1 | XP_023508766.1 | 2216 | 103 | |||
More Information
Set Membership: |
|

C11H17orf50
Genomic Map