Assay Details
| Gene Symbol: SNRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253840.1 | NP_001240769.1 | 2 - 3 | 142 | 141 | |
| XM_005604412.3 | XP_005604469.1 | 255 | 141 | |||
| XM_005604413.3 | XP_005604470.1 | 536 | 141 | |||
| XM_005604414.3 | XP_005604471.1 | 236 | 141 | |||
| XM_014734997.2 | XP_014590483.1 | 135 | 141 | |||
| XM_023625917.1 | XP_023481685.1 | 413 | 141 | |||
| GenBank mRNA | HQ889944.1 | - | 2 - 3 | 142 | 141 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001253840.1, XM_005604412.3, XM_005604413.3, XM_005604414.3, XM_014734997.2, XM_023625917.1 transcripts.
Set Membership: |
|

SNRPB2
Genomic Map