Assay Details
| Gene Symbol: SNRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253840.1 | NP_001240769.1 | 5 - 6 | 504 | 125 | |
| XM_005604412.3 | XP_005604469.1 | 617 | 125 | |||
| XM_005604413.3 | XP_005604470.1 | 898 | 125 | |||
| XM_005604414.3 | XP_005604471.1 | 598 | 125 | |||
| XM_014734997.2 | XP_014590483.1 | 497 | 125 | |||
| XM_023625917.1 | XP_023481685.1 | 775 | 125 | |||
| GenBank mRNA | HQ889944.1 | - | 5 - 6 | 504 | 125 | |
More Information
Set Membership: |
|

SNRPB2
Genomic Map