Assay Details
| Gene Symbol: PWWP3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256976.2 | NP_001243905.2 | 1 | 1819 | 100 | |
| XM_005614357.3 | XP_005614414.2 | 2898 | 100 | |||
| XM_005614358.3 | XP_005614415.2 | 2869 | 100 | |||
| XM_005614359.3 | XP_005614416.2 | 2910 | 100 | |||
| XM_005614360.3 | XP_005614417.2 | 2869 | 100 | |||
| XM_023633203.1 | XP_023488971.1 | 2892 | 100 | |||
| XM_023633204.1 | XP_023488972.1 | 2763 | 100 | |||
More Information
Set Membership: |
|

PWWP3B
Genomic Map