Assay Details
Gene Symbol: XIRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256977.2 | NP_001243906.2 | 1 | 5316 | 78 | |
XM_005600490.3 | XP_005600547.2 | 5489 | 78 | |||
XM_023619818.1 | XP_023475586.1 | 1707 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
