Assay Details
| Gene Symbol: ZNF324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257136.2 | NP_001244065.1 | 1 - 2 | 124 | 87 | |
| XM_005596102.3 | XP_005596159.1 | 515 | 87 | |||
| XM_005596103.3 | XP_005596160.1 | 513 | 87 | |||
More Information
Set Membership: |
|

ZNF324
Genomic Map