Assay Details
| Gene Symbol: ZNF324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257136.2 | NP_001244065.1 | 2 - 3 | 240 | 108 | |
| XM_005596103.3 | XP_005596160.1 | 629 | 108 | |||
| XM_023649410.1 | XP_023505178.1 | 197 | 108 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023649410.1 transcripts.
Set Membership: |
|

ZNF324
Genomic Map