Assay Details
| Gene Symbol: CALM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283082.1 | NP_001270011.1 | 5 - 6 | 480 | 78 | |
| GenBank mRNA | HQ889917.1 | - | 5 - 6 | 480 | 78 | |
More Information
Set Membership: |
|

CALM2
Genomic Map