Assay Details
| Gene Symbol: LOC100146423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283088.1 | NP_001270017.1 | 16 - 17 | 1735 | 70 | |
| XM_023621985.1 | XP_023477753.1 | 2191 | 70 | |||
| XM_023621987.1 | XP_023477755.1 | 2191 | 70 | |||
| XM_023621988.1 | XP_023477756.1 | 2190 | 70 | |||
| XM_023621989.1 | XP_023477757.1 | 2193 | 70 | |||
| XM_023621990.1 | XP_023477758.1 | 2184 | 70 | |||
More Information
Set Membership: |
|

LOC100146423
Genomic Map