Assay Details
| Gene Symbol: LOC100146423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283088.1 | NP_001270017.1 | 30 - 31 | 3441 | 96 | |
| XM_023621990.1 | XP_023477758.1 | 3893 | 96 | |||
More Information
Set Membership: |
|

LOC100146423
Genomic Map