Assay Details
| Gene Symbol: LOC100146423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283088.1 | NP_001270017.1 | 31 - 32 | 3557 | 74 | |
| XM_023621990.1 | XP_023477758.1 | 4009 | 74 | |||
More Information
Set Membership: |
|

LOC100146423
Genomic Map