Assay Details
| Gene Symbol: LOC100146423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283088.1 | NP_001270017.1 | 10 - 11 | 911 | 66 | |
| XM_023621985.1 | XP_023477753.1 | 1367 | 66 | |||
| XM_023621987.1 | XP_023477755.1 | 1367 | 66 | |||
| XM_023621988.1 | XP_023477756.1 | 1366 | 66 | |||
| XM_023621989.1 | XP_023477757.1 | 1369 | 66 | |||
| XM_023621990.1 | XP_023477758.1 | 1360 | 66 | |||
More Information
Set Membership: |
|

LOC100146423
Genomic Map