Assay Details
| Gene Symbol: C20H6orf89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301292.2 | NP_001288221.1 | 2 - 3 | 280 | 76 | |
| XM_005603856.3 | XP_005603913.1 | 446 | 76 | |||
More Information
Set Membership: |
|

C20H6orf89
Genomic Map