Assay Details
Gene Symbol: C20H6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301292.2 | NP_001288221.1 | 5 - 6 | 790 | 74 | |
XM_005603856.3 | XP_005603913.1 | 956 | 74 | |||
XM_014734213.2 | XP_014589699.1 | 595 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
