Assay Details
| Gene Symbol: C20H6orf89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301292.2 | NP_001288221.1 | 6 - 7 | 911 | 108 | |
| XM_005603856.3 | XP_005603913.1 | 1077 | 108 | |||
| XM_014734213.2 | XP_014589699.1 | 716 | 108 | |||
More Information
Set Membership: |
|

C20H6orf89
Genomic Map