Assay Details
| Gene Symbol: C16H3orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308948.1 | NP_001295877.1 | 2 - 3 | 592 | 104 | |
| XM_005600666.3 | XP_005600723.2 | 869 | 104 | |||
| XM_005600667.3 | XP_005600724.2 | 863 | 104 | |||
| XM_014731483.2 | XP_014586969.2 | 842 | 104 | |||
| GenBank mRNA | JL636408.1 | - | 2 - 3 | 591 | 104 | |
More Information
Set Membership: |
|

C16H3orf62
Genomic Map