Assay Details
Gene Symbol: SMIM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309469.1 | NP_001296398.1 | 2 - 3 | 112 | 76 | |
GenBank mRNA | CX596854.1 | - | 2 - 3 | 123 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001309469.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
