Assay Details
| Gene Symbol: LOC100051526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317252.1 | NP_001304181.1 | 1 - 2 | 36 | 96 | |
| XM_005609894.3 | XP_005609951.1 | 113 | 96 | |||
| XM_005609895.3 | XP_005609952.1 | 139 | 93 | |||
| GenBank mRNA | JN795139.1 | - | 1 - 2 | 36 | 96 | |
More Information
Set Membership: |
|

LOC100051526
Genomic Map