Assay Details
| Gene Symbol: KRT17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346193.1 | NP_001333122.1 | 6 - 7 | 1244 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT17| Gene Symbol: KRT17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346193.1 | NP_001333122.1 | 6 - 7 | 1244 | 83 | |
Set Membership: |
|