Assay Details
| Gene Symbol: POGLUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493228.4 | XP_001493278.1 | 1597 | 165 | ||
| GenBank mRNA | JL637886.1 | - | 1 - 2 | 216 | 165 | |
More Information
Set Membership: |
|

POGLUT2
Genomic Map