Assay Details
Gene Symbol: C4H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498834.4 | XP_001498884.1 | 148 | 87 | ||
XM_023639392.1 | XP_023495160.1 | 148 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001498834.4, XM_023639392.1 transcripts.
Set Membership: |
![]() ![]() |
