Assay Details
| Gene Symbol: CERT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599616.3 | XP_005599673.1 | 2180 | 72 | ||
| XM_005599617.3 | XP_005599674.1 | 2260 | 72 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CERT1| Gene Symbol: CERT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599616.3 | XP_005599673.1 | 2180 | 72 | ||
| XM_005599617.3 | XP_005599674.1 | 2260 | 72 | |||
Set Membership: |
|