Assay Details
Gene Symbol: SEPTIN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599751.3 | XP_005599808.1 | 465 | 105 | ||
XM_023618441.1 | XP_023474209.1 | 751 | 105 | |||
XM_023618443.1 | XP_023474211.1 | 751 | 105 | |||
XM_023618444.1 | XP_023474212.1 | 338 | 105 | |||
XM_023618445.1 | XP_023474213.1 | 751 | 105 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
