Assay Details
Gene Symbol: CYRIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023619492.1 | XP_023475260.1 | 223 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023619492.1 transcripts.
Set Membership: |
![]() ![]() |
