Assay Details
Gene Symbol: LOC100059872 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023620293.1 | XP_023476061.1 | 907 | 73 | ||
XM_023620294.1 | XP_023476062.1 | 908 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
