Assay Details
Gene Symbol: PRSS45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023620557.1 | XP_023476325.1 | 864 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
