Assay Details
Gene Symbol: LOC102147639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005601048.3 | XP_005601105.2 | 1110 | 108 | ||
XM_014731778.2 | XP_014587264.1 | 1116 | 108 | |||
XM_023620708.1 | XP_023476476.1 | 717 | 108 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
