Assay Details
| Gene Symbol: LOC102147639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005601048.3 | XP_005601105.2 | 1221 | 88 | ||
| XM_014731778.2 | XP_014587264.1 | 1227 | 88 | |||
| XM_023620708.1 | XP_023476476.1 | 828 | 88 | |||
More Information
Set Membership: |
|

LOC102147639
Genomic Map