Assay Details
Gene Symbol: ACP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495610.6 | XP_001495660.3 | 319 | 122 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
